missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDPCP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-58008
This item is not returnable.
View return policy
Description
WDPCP Polyclonal specifically detects WDPCP in Human samples. It is validated for Western Blot.
Specifications
| WDPCP | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Bardet-Biedl syndrome 15 protein, BBS15C2orf86FRITZ, chromosome 2 open reading frame 86, DKFZp686C12204, fritz, FRTZ, hFrtz, WD repeat containing planar cell polarity effector, WD repeat-containing and planar cell polarity effector protein, WD repeat-containing protein C2orf86 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51057 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| O95876-2 | |
| WDPCP | |
| Synthetic peptides corresponding to LOC51057(hypothetical protein LOC51057) The peptide sequence was selected from the N terminal of LOC51057. Peptide sequence LAQNKLCFIQFTKKMESSDVNKRLEKLSALDYKIFYYEIPGPINKTTERH. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Guinea pig: 100%; Human: 100%; Rat: 100%; Equine: 92%; Bovine: 84%; Rabbit: 78%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction