Learn More
Eppendorf Water Tank Holder for CellXpert CS220
Product Code. 30213532 Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
CellXpert CS220
Unit Size:
Each
Product Code. 30213532
Brand: Eppendorf 6740070098
Only null left
Add to basket
Add to basket
Accessory for CellXpert™ CS220 CO2 Incubator Shaker.
For a 5 L water tank
TRUSTED_SUSTAINABILITY
Specifications
Product Type rH-system
For Use With (Equipment) CellXpert CS220