missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Water Plate Count Prepared Agar, Bottled

Product Code. 11948419
Change view
Click to view available options
Fill Weight:
100 g +1.5 g
198 to 202 g
200 g + 2.5 g
248 to 252 g
Manufacturing Origin:
Germany
United Kingdom
Unit Size:
1 set
Pack of 10
Product Code. Fill Weight Manufacturing Origin unitSize
11948419 100 g +1.5 g United Kingdom Pack of 10
13205469 198 to 202 g United Kingdom Pack of 10
11958419 200 g + 2.5 g United Kingdom Pack of 10
15842287 248 to 252 g United Kingdom 1 set
13275439 100 g +1.5 g Germany Pack of 10
5 options
This item is not returnable. View return policy

Product Code. 11948419

Brand: Thermo Scientific™ BO0055M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Enumerate all culturable microorganisms from water using colony count technique with this nutritious and ISO compliant medium.

Enumerate all culturable microorganisms from water using colony count technique with Thermo Scientific™ Oxoid™ Water Plate Count Agar, ready prepared bottles. This nutritious, non-selective medium conforms to ISO 6222: 1999 (E) Water quality — Enumeration of culturable microorganisms — Colony count by inoculation in a nutrient agar culture medium1.

Waters of all kinds invariably contain a variety of microorganisms derived from various sources, such as soil and vegetation. Estimation of the overall numbers provide useful information for the assessment and surveillance of water quality. Separate counts are usually made of the microorganisms which are able to grow and form colonies on nutrient agar media at 36°C and 22°C.

Colony counts are useful for assessing the integrity of ground water sources and the efficiency of water treatment processes such as coagulation, filtration and disinfection and provide an indication of the cleanliness and integrity of the distribution system. They can also be used to assess the suitability of a water supply for the preparation of food and drink to avoid contaminating the product with spoilage organisms.

The main value of colony counts lies in the detection of changes from expected counts, based on frequent, long term monitoring. Any sudden increase in the count can be an early warning of serious pollution and a call for immediate investigation.1

Not all products are available for sale in all territories. Please inquire. Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY

Specifications

Color Straw
Content And Storage 2 to 25°C
Format 125 mL sirop bottle
Manufacturing Origin United Kingdom
Fill Weight 100 g +1.5 g
Packaging Type 10 sirop bottles in a box
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.