missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WASP Antibody (7B10E4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£187.00 - £438.00
Specifications
| Antigen | WASP |
|---|---|
| Clone | 7B10E4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18215630
|
Novus Biologicals
NBP2-52587 |
0.1 mg |
£438.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18669706
|
Novus Biologicals
NBP2-52587-0.025mg |
0.025 mg |
£187.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
WASP Monoclonal specifically detects WASP in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| WASP | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| Mouse | |
| Phospho Specific, Signal Transduction | |
| PBS with 0.05% Sodium Azide | |
| 7454 | |
| Purified recombinant fragment of human WAS (AA: 57-170) expressed in E. Coli. | |
| Primary | |
| 53 kDa |
| 7B10E4 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| IMD2THC, WASP, WiskTHC1, wiskthrombocytopenia 1 (X-linked) | |
| WAS | |
| IgG2a | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title