missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WASP Antibody (7B10E4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52587
This item is not returnable.
View return policy
Description
WASP Monoclonal specifically detects WASP in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| WASP | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin, CyTOF-ready | |
| IMD2THC, WASP, WiskTHC1, wiskthrombocytopenia 1 (X-linked) | |
| Mouse | |
| 53 kDa | |
| 0.1 mg | |
| Phospho Specific, Signal Transduction | |
| 7454 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| 7B10E4 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| WAS | |
| Purified recombinant fragment of human WAS (AA: 57-170) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction