missing translation for 'onlineSavingsMsg'
Learn More

WAPL/WAPAL Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen WAPL/WAPAL
Concentration 0.3 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16885964
missing translation for 'viewDocuments'
Proteintech
16370-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
16875964
missing translation for 'viewDocuments'
Proteintech
16370-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Transcriptional repressor which may play a role in development of the central nervous system (CNS).
Specifications

Specifications

WAPL/WAPAL
Western Blot
Unconjugated
Rabbit
Human, Mouse
Q65Z40, Q7Z5K2
218914, 23063
WAPL; WAPAL Fusion Protein Ag9280
Primary
-20°C
WAPL
0.3 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
FOE, Friend of EBNA2 protein, KIAA0261, WAPAL, WAPL
Wapl
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.