Learn More
WAPL/WAPAL Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16370-1-AP-150UL
 View more versions of this product
Product Code. 16875964
  • £349.00 / 150µL
Description
Transcriptional repressor which may play a role in development of the central nervous system (CNS).
Specifications
Specifications
WAPL/WAPAL
Polyclonal
Unconjugated
WAPL
FOE, Friend of EBNA2 protein, KIAA0261, WAPAL, WAPL
Rabbit
Antigen Affinity Chromatography
RUO
218914, 23063
-20°C
Liquid
Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q65Z40, Q7Z5K2
Wapl
WAPL; WAPAL Fusion Protein Ag9280
150 μL
Primary
Human, Mouse