missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ W.L. Nutrient Agar
Description
Differentially isolate and enumerate yeasts and bacteria in fermentation products such as beer and wine using W.L. Nutrient Agar.
Specifications
Specifications
| Color | Blue |
| Content And Storage | 2°C to 25°C |
| Format | 300 mL Sirop Bottle |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar |
| Fill Weight | 250 to 254 g |
| Packaging Type | 10 Sirop Bottles with screw cap closures in a box |
| Quantity | 10/Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?