missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VZV Glycoprotein B Antibody - (Oka vaccine), Novus Biologicals™
Rabbit Polyclonal Antibody
£154.00 - £302.00
Specifications
| Antigen | VZV Glycoprotein B |
|---|---|
| Dilution | Western Blot 1:2000-1:20000, ELISA 1:5000-1:10000 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30281521
|
Novus Biologicals
NBP3-43770-20ul |
20 μL |
£154.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30281520
|
Novus Biologicals
NBP3-43770-100ul |
100 μL |
£302.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VZV Glycoprotein B Polyclonal antibody specifically detects VZV Glycoprotein B in Virus samples. It is validated for ELISA,Western BlotSpecifications
| VZV Glycoprotein B | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| chicken pox, gB, gp140-62, gpII, ORF 31, varicella zoster, VZV | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:2000-1:20000, ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 μm filtered solution in PBS | |
| Produced in rabbits immunized with purified, recombinant Varicella Zoster Virus Glycoprotein B (Oka vaccine) (Uniprot#: Q4JR05) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title