missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VZV Glycoprotein B Antibody - (Oka vaccine), Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-43770-20ul
This item is not returnable.
View return policy
Description
VZV Glycoprotein B Polyclonal antibody specifically detects VZV Glycoprotein B in Virus samples. It is validated for ELISA,Western Blot
Specifications
| VZV Glycoprotein B | |
| Polyclonal | |
| Western Blot 1:2000-1:20000, ELISA 1:5000-1:10000 | |
| chicken pox, gB, gp140-62, gpII, ORF 31, varicella zoster, VZV | |
| Produced in rabbits immunized with purified, recombinant Varicella Zoster Virus Glycoprotein B (Oka vaccine) (Uniprot#: Q4JR05) | |
| 20 μL | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction