missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VPS34 Recombinant Rabbit Monoclonal Antibody (24GB1150)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£172.00 - £488.00

Specifications

Antigen VPS34
Clone 24GB1150
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30274788
GREENER_CHOICE
View Documents
Invitrogen™
MA557604
100 μL
£488.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30275992
View Documents
Invitrogen™
MA557605
20 μL
£172.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

VPS34 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

PIK3C3 (hVPS34) is the sole class III member of the PI3 Kinase family. PIK3C3 phosphorylates phosphatidylinositol (PI) to produce phosphatidylinositol 3-phosphate [PI(3)P], which recruits downstream effectors containing FYVE and Phox domains to membranes. PIK3C3 is involved in coupling nutrient signaling to mTOR and also plays a role in autophagy and intracellular trafficking. Involved in regulation of degradative endocytic trafficking and required for the abcission step in cytokinesis, probably in the context of PI3KC3-C2. Involved in the transport of lysosomal enzyme precursors to lysosomes. Required for transport from early to late endosomes.
Specifications

Specifications

VPS34
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse
Pik3c3
5330434F23Rik; catalytic phosphatidylinositol 3-kinase 3; catalytic subunit type 3 Antibody; class 3 phosphoinositide-3-kinase; Class III PI 3-kinase; hVps34; hVps34 Antibody; OTTMUSP00000023341; OTTMUSP00000023350; phosphatidylinositol 3-kinase; Phosphatidylinositol 3-kinase catalytic subunit type 3; phosphatidylinositol 3-kinase p100 subunit; phosphatidylinositol 3-kinase, catalytic subunit type 3; Phosphoinositide-3-kinase class 3; phosphoinositide-3-kinase, class 3; PI3K type 3; PI-3K Vps34p; pi3kc3; PI3-kinase type 3; Pik3c3; PIK3C3 Antibody; PtdIns-3-kinase type 3; VPS34
Pik3c3
Primary
Affinity chromatography
24GB1150
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q6PF93, Q8NEB9
225326, 5289
A synthesized peptide derived from human PI 3 Kinase Class 3 (aa 800-887).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.