Learn More
Invitrogen™ VPS34 Recombinant Rabbit Monoclonal Antibody (24GB1150)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557604
 View more versions of this product
Product Code. 30274788
  • £488.00 / 100µL
Description
VPS34 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
PIK3C3 (hVPS34) is the sole class III member of the PI3 Kinase family. PIK3C3 phosphorylates phosphatidylinositol (PI) to produce phosphatidylinositol 3-phosphate [PI(3)P], which recruits downstream effectors containing FYVE and Phox domains to membranes. PIK3C3 is involved in coupling nutrient signaling to mTOR and also plays a role in autophagy and intracellular trafficking. Involved in regulation of degradative endocytic trafficking and required for the abcission step in cytokinesis, probably in the context of PI3KC3-C2. Involved in the transport of lysosomal enzyme precursors to lysosomes. Required for transport from early to late endosomes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VPS34
Recombinant Monoclonal
Unconjugated
Pik3c3
5330434F23Rik; catalytic phosphatidylinositol 3-kinase 3; catalytic subunit type 3 Antibody; class 3 phosphoinositide-3-kinase; Class III PI 3-kinase; hVps34; hVps34 Antibody; OTTMUSP00000023341; OTTMUSP00000023350; phosphatidylinositol 3-kinase; Phosphatidylinositol 3-kinase catalytic subunit type 3; phosphatidylinositol 3-kinase p100 subunit; phosphatidylinositol 3-kinase, catalytic subunit type 3; Phosphoinositide-3-kinase class 3; phosphoinositide-3-kinase, class 3; PI3K type 3; PI-3K Vps34p; pi3kc3; PI3-kinase type 3; Pik3c3; PIK3C3 Antibody; PtdIns-3-kinase type 3; VPS34
Rabbit
Affinity chromatography
RUO
225326, 5289
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1150
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q6PF93, Q8NEB9
Pik3c3
A synthesized peptide derived from human PI 3 Kinase Class 3 (aa 800-887).
100 μL
Primary
Human, Mouse