missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS29 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | VPS29 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230394
|
Novus Biologicals
NBP3-35309-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231326
|
Novus Biologicals
NBP3-35309-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VPS29 Polyclonal antibody specifically detects VPS29 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ImmunofluorescenceSpecifications
| VPS29 | |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Protein Phosphatase | |
| PBS (pH 7.3), 50% glycerol | |
| 51699 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DC15, DC7, DKFZp564F0223, EC 3.1.3.3, FLJ20492, hVPS29, PEP11, PEP11 homolog, retromer protein, vacuolar protein sorting 29 (yeast homolog), vacuolar protein sorting 29 (yeast), vacuolar protein sorting 29 homolog (S. cerevisiae), vacuolar protein sorting-associated protein 29, vacuolar sorting protein VPS29/PEP11, Vesicle protein sorting 29, x 007 protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-186 of human VPS29 (NP_476528.1).,, Sequence:, MAGHRLVLVLGDLHIPHRCNSLPAKFKKLLVPGKIQHILCTGNLCTKESYDYLKTLAGDVHIVRGDFDENLNYPEQKVVTVGQFKIGLIHGHQVIPWGDMASLALLQRQFDVDILISGHTHKFEAFEHENKFYINPGSATGAYNALETNIIPSFVLMDIQASTVVTYVYQLIGDDVKVERIEYKKP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title