missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS29 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35309-20ul
This item is not returnable.
View return policy
Description
VPS29 Polyclonal antibody specifically detects VPS29 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| VPS29 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| DC15, DC7, DKFZp564F0223, EC 3.1.3.3, FLJ20492, hVPS29, PEP11, PEP11 homolog, retromer protein, vacuolar protein sorting 29 (yeast homolog), vacuolar protein sorting 29 (yeast), vacuolar protein sorting 29 homolog (S. cerevisiae), vacuolar protein sorting-associated protein 29, vacuolar sorting protein VPS29/PEP11, Vesicle protein sorting 29, x 007 protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-186 of human VPS29 (NP_476528.1).,, Sequence:, MAGHRLVLVLGDLHIPHRCNSLPAKFKKLLVPGKIQHILCTGNLCTKESYDYLKTLAGDVHIVRGDFDENLNYPEQKVVTVGQFKIGLIHGHQVIPWGDMASLALLQRQFDVDILISGHTHKFEAFEHENKFYINPGSATGAYNALETNIIPSFVLMDIQASTVVTYVYQLIGDDVKVERIEYKKP | |
| 20 μL | |
| Autophagy, Protein Phosphatase | |
| 51699 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction