missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£130.00 - £339.00
Specifications
Antigen | VPS15 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Western Blot, Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16877894
|
Proteintech
17894-1-AP-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16867894
|
Proteintech
17894-1-AP-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains.Specifications
VPS15 | |
Western Blot, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
P0C0R5, Q8VD65, Q99570 | |
30849, 363131, 75669 | |
VPS15 Fusion Protein Ag12221 | |
Primary | |
-20°C | |
PIK3R4 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
p150, PI3 kinase p150 subunit, PI3K p150, PI3-kinase regulatory subunit 4, PIK3R4, VPS15 | |
PIK3R4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |