missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17894-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains.Specifications
VPS15 | |
Polyclonal | |
Unconjugated | |
PIK3R4 | |
p150, PI3 kinase p150 subunit, PI3K p150, PI3-kinase regulatory subunit 4, PIK3R4, VPS15 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
30849, 363131, 75669 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P0C0R5, Q8VD65, Q99570 | |
PIK3R4 | |
VPS15 Fusion Protein Ag12221 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |