missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VprBP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£167.00 - £310.00
Specifications
| Antigen | VprBP |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2-4ug/ml∼, Flow Cytometry 0.2ug/10^6cells |
| Applications | Western Blot, Flow Cytometry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18113752
|
Novus Biologicals
NBP2-27203SS |
0.025 mg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18199613
|
Novus Biologicals
NBP2-27203 |
0.1 mg |
£310.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VprBP Polyclonal specifically detects VprBP in Human, Mouse, Rat, Porcine, Chicken, Chinese Hamster, Equine, Feline, Guinea Pig, Opossum, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry.Specifications
| VprBP | |
| Western Blot 2-4ug/ml∼, Flow Cytometry 0.2ug/10^6cells | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| DCAF1DDB1 and CUL4 associated factor 1, DDB1- and CUL4-associated factor 1, HIV-1 Vpr-binding protein, KIAA0800protein VPRBP, MGC102804, RIP, Vpr (HIV-1) binding protein, Vpr-binding protein, VprBP, Vpr-interacting protein | |
| VPRBP | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in human, mouse, rat, european pole-cat, pig, orangutan, rabbit, white-cheeked gibbon, opossum, wild turkey, african bush elephant, chicken, horse, Chinese hamster, guinea pig, and panda, 94% in cow, and platypus, and 88% homolgous in western clawed frog.There are numerous isoforms including 1505, 1504, 1076 and 783 amino acids. |
| 0.5 mg/mL | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat, Pig, Chicken, Chinese Hamster, Equine, Feline, Guinea Pig, Opossum, Other, Primate, Rabbit | |
| Q9Y4B6 | |
| 9730 | |
| A portion of amino acids 935-985 from human VPRBP was used as the immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title