missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VprBP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27203
This item is not returnable.
View return policy
Description
VprBP Polyclonal specifically detects VprBP in Human, Mouse, Rat, Porcine, Chicken, Chinese Hamster, Equine, Feline, Guinea Pig, Opossum, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| VprBP | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| DCAF1DDB1 and CUL4 associated factor 1, DDB1- and CUL4-associated factor 1, HIV-1 Vpr-binding protein, KIAA0800protein VPRBP, MGC102804, RIP, Vpr (HIV-1) binding protein, Vpr-binding protein, VprBP, Vpr-interacting protein | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 9730 | |
| Human, Mouse, Rat, Pig, Chicken, Chinese Hamster, Equine, Feline, Guinea Pig, Opossum, Other, Primate, Rabbit | |
| Purified |
| Western Blot, Flow Cytometry | |
| 0.5 mg/mL | |
| Western Blot 2-4ug/ml∼, Flow Cytometry 0.2ug/10^6cells | |
| Q9Y4B6 | |
| VPRBP | |
| A portion of amino acids 935-985 from human VPRBP was used as the immunogen for this antibody. | |
| 0.1 mg | |
| Primary | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in human, mouse, rat, european pole-cat, pig, orangutan, rabbit, white-cheeked gibbon, opossum, wild turkey, african bush elephant, chicken, horse, Chinese hamster, guinea pig, and panda, 94% in cow, and platypus, and 88% homolgous in western clawed frog.There are numerous isoforms including 1505, 1504, 1076 and 783 amino acids. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction