missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VNN1 Monoclonal Antibody (C2), Invitrogen™

Mouse Monoclonal Antibody
£161.00 - £369.00
Specifications
| Antigen | VNN1 |
|---|---|
| Clone | C2 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17949952
|
Invitrogen™
MA551160 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17980573
![]() |
Invitrogen™
MA551159 |
200 μL |
£369.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hematopoietic precursor cells migrate to the thymus, where they differentiate into mature T lymphocytes. GPI-anchored vanin-1 protein regulates the late adhesion steps of thymus homing of bone marrow precursor cells. Vanin-1 is ubiquitously expressed as a pantetheinase enzyme and catalyzes the hydrolysis of pantetheine for vitamin B5 recycling. The hydrolytic activity of vanin-1 generates the potent antioxidant cysteamine as a metabolite. As a membrane bound pantetheinase, vanin-1 provides the main source of cysteamine under normal physiological conditions. In mice, vanin-1 is expressed specifically in male Sertoli cells of the developing testis, where it aids in cell migration.Specifications
| VNN1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| HDLCQ8; Pantetheinase; pantetheine hydrolase; Tiff66; V-1; vanin 1; Vanin1; vanin-1; vannin 1; vascular non-inflammatory molecule 1; VNN1 | |
| VNN1 | |
| IgG1 κ | |
| Protein A/G | |
| VNN1 |
| C2 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Pig | |
| O95497, Q9BDJ5 | |
| 397246, 8876 | |
| Recombinant Human VNN1 protein, Gln22-Gly491 (Accession #O95497). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title