missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VNN1 Monoclonal Antibody (C2), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA551160

 View more versions of this product

Product Code. 17949952

  • £161.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Hematopoietic precursor cells migrate to the thymus, where they differentiate into mature T lymphocytes. GPI-anchored vanin-1 protein regulates the late adhesion steps of thymus homing of bone marrow precursor cells. Vanin-1 is ubiquitously expressed as a pantetheinase enzyme and catalyzes the hydrolysis of pantetheine for vitamin B5 recycling. The hydrolytic activity of vanin-1 generates the potent antioxidant cysteamine as a metabolite. As a membrane bound pantetheinase, vanin-1 provides the main source of cysteamine under normal physiological conditions. In mice, vanin-1 is expressed specifically in male Sertoli cells of the developing testis, where it aids in cell migration.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

VNN1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
O95497, Q9BDJ5
VNN1
Recombinant Human VNN1 protein, Gln22-Gly491 (Accession #O95497).
20 μL
Primary
Human, Pig
Antibody
IgG1 κ
Western Blot
C2
Unconjugated
VNN1
HDLCQ8; Pantetheinase; pantetheine hydrolase; Tiff66; V-1; vanin 1; Vanin1; vanin-1; vannin 1; vascular non-inflammatory molecule 1; VNN1
Mouse
Protein A/G
RUO
397246, 8876
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.