missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Vivapure™ Virus Purification Kits
BIOPHARMA

Product Code. p-8006484 Shop All Sartorius Products
Click to view available options
Sample Size:
20 mL
500 mL
2 x 100 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11710855

Brand: Sartorius™ VSAVPQ022

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The adenovirus purification kits are ready to use and contain all of the buffers and devices you need for quantitative yield virus purification in just a few hours.

  • Membrane adsorber technology
  • Fast and safe virus purification
  • Ready to use - kits contain all buffers and devices you need
TRUSTED_SUSTAINABILITY

Specifications

Endotoxin Concentration <0.025 EU/mL
Ligand Type Quaternary ammonium
Includes 6 x AdenoPACK Maxi spin columns with Sartobind™ ion exchange membrane, clarification and concentration units, all buffers excluding Benzonase™
Sample Size 20 mL
Test Time 1 hr.
Purification Target Upto 1 x 1012 adenosvirus type 5 particles from 20 mL cell culture
No. of Preps 6 x 20 mL
Sample Type Cell culture
For Use With (Application) Adeno virus purification type 5
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.