missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ Vivapure™ Virus Purification Kits
Shop All Sartorius Products
Click to view available options
Sample Size:
20 mL
500 mL
2 x 100 mL
Unit Size:
Each
Description
- Membrane adsorber technology
- Fast and safe virus purification
- Ready to use - kits contain all buffers and devices you need
Specifications
Specifications
| Ligand Type | Quaternary ammonium |
| Includes | 2 x Adenopack 100 syringe filter units with Sartobind™, ion exchange membrane, all buffers and other accessories; without DNA-digesting enzyme |
| Sample Size | 2 x 100 mL |
| Test Time | 2 hours |
| Purification Target | Upto 1 x 1013 Adenovirus type 5 particles from 500 mL of cell culture. |
| Elution Volume Max | 200 mL |
| Elution Volume Min | 20 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction