missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Vivapure™ Virus Purification Kits
BIOPHARMA

Product Code. 11710855 Shop All Sartorius Products
Change view
Click to view available options
Sample Size:
20 mL
500 mL
2 x 100 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sample Size unitSize
11710855 20 mL Each
11750855 500 mL Each
11730855 2 x 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11710855 Supplier Sartorius™ Supplier No. VSAVPQ022

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The adenovirus purification kits are ready to use and contain all of the buffers and devices you need for quantitative yield virus purification in just a few hours.

  • Membrane adsorber technology
  • Fast and safe virus purification
  • Ready to use - kits contain all buffers and devices you need
TRUSTED_SUSTAINABILITY

Specifications

Endotoxin Concentration <0.025 EU/mL
Ligand Type Quaternary ammonium
Includes 6 x AdenoPACK Maxi spin columns with Sartobind™ ion exchange membrane, clarification and concentration units, all buffers excluding Benzonase™
Sample Size 20 mL
Test Time 1 hr.
Purification Target Upto 1 x 1012 adenosvirus type 5 particles from 20 mL cell culture
No. of Preps 6 x 20 mL
Sample Type Cell culture
For Use With (Application) Adeno virus purification type 5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.