missing translation for 'onlineSavingsMsg'
Learn More

Isofield™ Vista Sterile Cleanroom Coverall with Hood - Grade A/B Compatible
GREENER_CHOICEBIOPHARMA

Product Code. 30178859
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 15
Pack of 20
This item is not returnable. View return policy

Product Code. 30178859

Brand: Isofield™ 80011L

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The Vista sterile coverall with postbox hood for complete head coverage is aseptically folded, validated sterile, and tested against chemo to ASTM F 739. The anti-static, low linting, PPE Cat III Type 5/6 Vista provides essential protection in Grade A/B environments for all cleanroom activities.

  • Sterile coverall with Innovative Postbox hood design
  • Taped zip flap, elasticated hood, waist, wrists, and ankles
  • Aseptically Folded
  • Class 1 for Cleanliness, static-dissipative and tested against Chemotherapy
  • Microporous Polypropylene / Polyethylene
  • 60 gsm, Low-linting robust fabric
  • Excellent low-hazard liquid spray and fine particulates protection

Technical and Documentation Compliance

  • COA, COC, COI Available for download online
  • QR Code on cartons allows for immediate Document retrieval
  • Repellency to chemicals and Resistance to Penetration by chemicals EN 14325:2004, EN ISO 6530:2005
  • Abrasion Resistance EN 14325:2004, EN ISO 12947-2:2016
  • Flex Cracking Resistance EN 14325:2004, EN ISO 7854:1997, Method B
  • Trapezoidal Tear Resistance EN 14325:2004
  • Machine Direction and Cross Direction EN ISO 9073-4:1997
  • Tensile Strength, Cross Direction EN 14325:2004, EN ISO 13934-1:2013
  • Machine Direction EN ISO 13934-1:2013
  • Puncture Resistance EN 14325:2004, EN 863:1995
  • Seam Strength EN 14325:2004 [5.5]
  • Inward Leakage of aerosols and fine particles into suits EN ISO 13982- 2:2004
  • IL 82/90 EN ISO 13982- 1:2004
  • IL 8/10 (Type 5)
  • Resistance to penetration by a spray of liquid (Spray Test) EN 13034:2015 + A1:2009, ISO 17491-4:2008, (Type 6)
  • Permeation of Liquids and Gases through Protective Clothing under conditions of Continuous Contact ASTM F739-20
  • Sterility Validation EN ISO 11137 Part 2 : 2015
  • Particulate Cleanliness – Helmke Drum Test : Class 1
  • Surface Resistivity ASTM D257

Specifications

Product Type Cleanroom Coveralls
Size Large
Gender Unisex
Material Polyethylene, Non-woven Polypropylene Laminate
Color White
Disposable Single-use
Waist Style Elastic at Back
Ankle Style Open
Cuff Style Covered Elastic
Neck Style Hooded
Hood Type Attached, Open Face
Closure Type Zipper
Seam Type Bound
Sterility Sterile
Resistance Chemicals, Anti-static, Liquid, Particle
No. of Pockets None
Cleanroom Class Class 100 (ISO 5)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.