Learn More
VISTA Monoclonal Antibody (MIH64), Super Bright™ 436, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 62-1083-80
Description
Description: This MIH64 monoclonal antibody recognizes mouse VISTA also known as B7-H5 or PD-H1. This MIH64 antibody will work in flow cytometry on both native and paraformaldehyde-fixed cells. Applications Reported: This MIH64 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MIH64 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 1 μg/mL per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
VISTA Monoclonal antibody specifically detects VISTA in Mouse samples. It is validated for Flow Cytometry
Specifications
| VISTA | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Q9D659 | |
| VSIR | |
| Affinity chromatography | |
| RUO | |
| 74048 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| MIH64 | |
| Super Bright 436 | |
| VSIR | |
| B7H5; B7-H5; C10orf54; DD1alpha; Dies1; GI24; PD-1H; PD-H1; Platelet receptor Gi24; PP2135; SISP1; Sisp-1; Stress-induced secreted protein-1; UNQ730/PRO1412; VISTA; V-set domain-containing immunoregulatory receptor; V-set immunoregulatory receptor; Vsir; V-type immunoglobulin domain-containing suppressor of T-cell activation | |
| Rat | |
| 25 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.