missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Visocolor™ Eco Colorimetric and Titrimetric Test Kits
Shop All Macherey-Nagel ProductsDescription
- Colorimetric and titrimetric test kits
- Avoids hazardous substances wherever possible
Specifications
Specifications
| For Use With (Application) | To determine ammonia in water |
| Includes | 30mL NH4-1, 2.5 g NH4-2, 6mL NH4-3, 70mm measuring spoon, 2 screw-plug measuring glasses, slide comparator, color chart, 1m plastic syringe, instructions for use |
| Quantity | 40 g |
| Range | 0 · 0.5 · 1 · 2 · 3 · 5 · 7 · 10 · 15mg/L NH4+ |
| Sufficient For | 50 Tests |
| Test Method | Colorimetric method |
| Type | Visocolor Eco Ammonium 15 |
Safety and Handling
Signal Word
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Flammable liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H226-Flammable liquid and vapour.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?