missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Visocolor™ HE Titration Test Kit Refill Pack

Product Code. 10047700 Shop All Macherey-Nagel Products
Change view
Click to view available options
No. of Tests:
300
Unit Size:
143g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Tests unitSize
10047700 300 143g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10047700 Supplier Macherey-Nagel™ Supplier No. 915204

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Replace consumed chemicals from test kits. Macherey-Nagel™ Visocolor™ HE Titration Test Kit Refill Packs are reagents used with HE tests to determine even the lowest limiting values. The kits use longer measuring tubes and larger sample volumes for the highest sensitivity and accuracy.

Visocolor™ HE test kits provide highly precise analysis via exact dosing of the reagents, as well as by compensating for turbidity and color. The sensitivity of Visocolor™ HE is 10 to 100 times higher compared to other Visocolor tests. Each test kit is economically priced and provides convenient handling.

  • High sensitive volumetric analysis via visual evaluation
  • Narrow syringe with gradation marks for easy, accurate readings right on the barrel
  • Gradations show equivalent analyte level based on titrant required for color change
  • Sharper color change due to separated dropping reagents
  • Refill packs and replacement titration syringes available
  • Single reagent bottles with clear dosing instructions
  • Delivered in a robust box with plastic inlay and easy-to-understand instructions

Specifications

Analytes Chloride
Color Blue to Yellow to Violet
Description Chloride CL 500
Format Visocolor™ HE Test Kit
For Use With (Application) Chloride determination
For Use With (Equipment) Visocolor™ HE Test Kit for Chloride CL 500 (915 004)
Graduations 5mg/L gradation mark
Includes Indicator, reagent and titration solutions
No. of Tests 300
Quantity 1 Each
Range 5 to 500mg/L Cl-
Test Method Mercurimetric titration
Type Reagent refill pack
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • visocolour HE chloride CL 500
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H312-Harmful in contact with skin.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 1,5-diphenylccarbazone, ethanol, nitricacid, mercury (II) nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.