missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Visocolor™ ECO Copper Colorimetric Test Kit

Product Code. 10449472 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
1 test
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10449472 1 Each 1 test
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10449472 Supplier Macherey-Nagel™ Supplier No. 931037

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

For Use With (Application) In weakly alkaline medium copper(II) ions react with cuprizone forming a blue color complex
Quantity 1 Each
Range 0 to 1.5mg/L Cu
Sufficient For 100 Determinations
Type ECO Copper Colorimetric Test Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.