missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral Dengue Virus 4 NS1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£156.00 - £345.00
Specifications
| Antigen | Dengue virus 4 NS1 |
|---|---|
| Clone | 971314 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18189380
|
R&D Systems
MAB9444-SP |
25 ug |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18758648
![]() |
R&D Systems
MAB9444-100 |
100 μg |
£345.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dengue Virus 4 NS1 Monoclonal specifically detects Dengue Virus 4 NS1 in Viral samples. It is validated for Western Blot.Specifications
| Dengue virus 4 NS1 | |
| 1.0 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Virus | |
| DEN-4, Dengue NS1, Dengue pan, Dengue virus Serotype 4, Dengue virus Type 4, DENV4, DENV4-NS1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects recombinant Dengue Virus 4 NS1 in direct and capture ELISAs and Western blots. In direct ELISAs and Western Blots, no cross-reactivity with recombinant Dengue Virus 1, 2 and 3 NS1 proteins was observed. |
| 971314 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| NP_740318 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
RUO
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title