Learn More
Viral Dengue Virus 4 NS1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB9444-SP
 View more versions of this product
Product Code. 18189380
  • £156.00 / 25µg
Description
Dengue Virus 4 NS1 Monoclonal specifically detects Dengue Virus 4 NS1 in Viral samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dengue virus 4 NS1
Monoclonal
1.0 mg/mL
Western Blot 1 ug/mL
NP_740318
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Detects recombinant Dengue Virus 4 NS1 in direct and capture ELISAs and Western blots. In direct ELISAs and Western Blots, no cross-reactivity with recombinant Dengue Virus 1, 2 and 3 NS1 proteins was observed.
Virus
Purified
Western Blot
971314
Unconjugated
Lyophilized from a 0.2 um filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. with No Preservative
DEN-4, Dengue NS1, Dengue pan, Dengue virus Serotype 4, Dengue virus Type 4, DENV4, DENV4-NS1
Human embryonic kidney cell line HEK293-derived recombinant Dengue Virus 4 NS1 protein Asp1-Ala352 Accession No. NP_740318
25 ug
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b