missing translation for 'onlineSavingsMsg'
Learn More

Vinylcyclopentane, 99%

Product Code. 11306679
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
missing translation for 'unitSize'
1g
5g
This item is not returnable. View return policy

Product Code. 11306679

missing translation for 'mfr': Thermo Scientific Alfa Aesar H26230.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3742-34-5 | C7H12 | 96.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3742-34-5
Molecular Formula C7H12
Molecular Weight (g/mol) 96.17
MDL Number MFCD00001386
InChI Key BEFDCLMNVWHSGT-UHFFFAOYSA-N
Synonym vinylcyclopentane, cyclopentane, ethenyl, cyclopentane, vinyl, cyclopentyl-ethene, cyclopentylethylene, cyclopentane,ethenyl, acmc-209iss
PubChem CID 77345
IUPAC Name ethenylcyclopentane
SMILES C=CC1CCCC1

Specifications

Density 0.704
Boiling Point 97°C
Flash Point 10°C (50°F)
Refractive Index 1.436
Quantity 1 g
UN Number UN3295
Formula Weight 96.17
Percent Purity 99%
Chemical Name or Material Vinylcyclopentane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.