CAS RN 3742-34-5
CAS RN 3742-34-5
IUPAC Name:
ethenylcyclopentane
Synonyms:
ethenylcyclopentane
Molecular Weight (g/mol):
96.17
Molecular Formula:
C7H12
InChi Key:
BEFDCLMNVWHSGT-UHFFFAOYSA-N
SMILES:
C=CC1CCCC1
1
–
2
of
2
results
1
Vinylcyclopentane, 99%
CAS: 3742-34-5 Molecular Formula: C7H12 Molecular Weight (g/mol): 96.17 MDL Number: MFCD00001386 InChI Key: BEFDCLMNVWHSGT-UHFFFAOYSA-N Synonym: vinylcyclopentane,cyclopentane, ethenyl,cyclopentane, vinyl,cyclopentyl-ethene,cyclopentylethylene,cyclopentane,ethenyl,acmc-209iss PubChem CID: 77345 IUPAC Name: ethenylcyclopentane SMILES: C=CC1CCCC1