missing translation for 'onlineSavingsMsg'
Learn More

Vinculin Antibody (VIN-11-5), Novus Biologicals™

Mouse Monoclonal Antibody

£170.00

Specifications

Antigen Vinculin
Clone VIN-11-5
Dilution Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:50, Microarray
Applications Western Blot
Classification Monoclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18575684
View Documents
Novus Biologicals
NB120-11193-0.025ml
0.025 mL
£170.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Vinculin Monoclonal specifically detects Vinculin in Human, Mouse, Bovine, Chicken samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Microarray.
Specifications

Specifications

Vinculin
Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:50, Microarray
Monoclonal
Ascites
RUO
Human
P12003
7414
smooth muscle vinculin from chicken gizzard.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
116 kDa
VIN-11-5
Western Blot
Unconjugated
Mouse
Cellular Markers, Cytoskeleton Markers
Ascites with 15mM Sodium Azide
CMD1W, CMH15, Metavinculin, MVCL, vinculin
VCL
IgG1
Unpurified
The reacts best with cultured chicken fibroblasts. Labeling also may be obtained with bovine, human, or mouse cells.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.