missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinculin Antibody (VIN-11-5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-11193-0.025ml
This item is not returnable.
View return policy
Description
Vinculin Monoclonal specifically detects Vinculin in Human, Mouse, Bovine, Chicken samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Microarray.
Specifications
| Vinculin | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 15mM Sodium Azide | |
| CMD1W, CMH15, Metavinculin, MVCL, vinculin | |
| Mouse | |
| 116 kDa | |
| 0.025 mL | |
| Cellular Markers, Cytoskeleton Markers | |
| 7414 | |
| Human | |
| Ascites |
| Western Blot | |
| VIN-11-5 | |
| Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:50, Microarray | |
| P12003 | |
| VCL | |
| smooth muscle vinculin from chicken gizzard. | |
| Unpurified | |
| RUO | |
| Primary | |
| The reacts best with cultured chicken fibroblasts. Labeling also may be obtained with bovine, human, or mouse cells. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction