missing translation for 'onlineSavingsMsg'
Learn More

Vimentin, p Ser72 Antibody (SR1947), Novus Biologicals™

Rabbit Monoclonal Antibody

£257.00 - £341.00

Specifications

Antigen Vimentin
Clone SR1947
Dilution Western Blot 1:500-1:2000, Immunoprecipitation 1:50
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18679384
View Documents
Novus Biologicals
NBP3-21510-50ul
50 μg
£257.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18639334
View Documents
Novus Biologicals
NBP3-21510-100ul
100 μg
£341.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Vimentin Monoclonal antibody specifically detects Vimentin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications

Specifications

Vimentin
Western Blot 1:500-1:2000, Immunoprecipitation 1:50
Monoclonal
Purified
RUO
Human, Mouse, Rat
epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin
A synthesized peptide derived from human phospho-Vimentin [Ser72] (Uniprot #: P08670)
Primary
Store at -20 degrees C. Avoid freeze/thaw cycles.
SR1947
Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
7431
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.