missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vimentin, p Ser72 Antibody (SR1947), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21510-50ul
This item is not returnable.
View return policy
Description
Vimentin Monoclonal antibody specifically detects Vimentin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Vimentin | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SR1947 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin | |
| A synthesized peptide derived from human phospho-Vimentin [Ser72] (Uniprot #: P08670) | |
| 50 μg | |
| Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells | |
| 7431 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction