missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vimentin Ab-2 Mouse Monoclonal Antibody, Epredia™
Epredia™ Vimentin Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.
£92.55 - £1375.00
Specifications
Antigen | Vimentin Ab-2 |
---|---|
Clone | V9 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Host Species | Mouse |
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
12679506
|
Epredia
MS-129-B0 |
Concentrated | Biotin | 100 μL |
£155.00
100µL |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12689506
|
Epredia
MS-129-B1 |
Concentrated | Biotin | 500 μL |
£379.00
500µL |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12639516
|
Epredia
MS-129-PABX |
Concentrated | Unconjugated | 200 μL |
£1375.00
200µL |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12619516
|
Epredia
MS-129-P1 |
Concentrated | Unconjugated | 500 μL |
£283.00
500µL |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12699506
|
Epredia
MS-129-P |
Concentrated | Unconjugated | 1 mL |
£351.00
1mL |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12679516
|
Epredia
MS-129-RQ |
Ready-to-Use | Unconjugated | 12 mL |
£247.00
12mL |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12609516
|
Epredia
MS-129-P0 |
Concentrated | Unconjugated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12629516
|
Epredia
MS-129-P1ABX |
Concentrated | Unconjugated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12669516
|
Epredia
MS-129-R7 |
Prediluted | Unconjugated | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12669506
|
Epredia
MS-129-B |
Concentrated | Biotin | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Vimentin is the main intermediate filament protein in mesenchymal cells and is therefore of value in the differential diagnosis of undifferentiated neoplasms.Host Species: Mouse
Clone: V9
Isotype: IgG1/κ
Species Reactivity: Rat, Horse, Chicken, Cow, Cat, Dog, Human and Pig. Others not known.
Epitope: Not determined
Immunogen: Purified vimentin from pig eye lens
Molecular Weight: 57-60kDa
Positive Control: Sarcomas
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Vimentin Ab-2 | |
Immunohistochemistry (Paraffin) | |
Mouse | |
Cytoskeletal Protein | |
Primary |
V9 | |
Monoclonal | |
IVD | |
Purified vimentin from pig eye lens |