Learn More
Vimentin Ab-2 Mouse Monoclonal Antibody, Epredia™
Epredia™ Vimentin Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.
Brand: Epredia MS-129-P1
Description
Vimentin is the main intermediate filament protein in mesenchymal cells and is therefore of value in the differential diagnosis of undifferentiated neoplasms.Host Species: Mouse
Clone: V9
Isotype: IgG1/κ
Species Reactivity: Rat, Horse, Chicken, Cow, Cat, Dog, Human and Pig. Others not known.
Epitope: Not determined
Immunogen: Purified vimentin from pig eye lens
Molecular Weight: 57-60kDa
Positive Control: Sarcomas
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| Vimentin Ab-2 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 500 μL | |
| Cytoskeletal Protein | |
| Bovine, Canine, Chicken, Equine, Feline, Gerbil, Hamster, Human, Mouse, Non-human Primate, Pig, Rabbit, Rat | |
| IgG2a κ |
| Immunohistochemistry (Paraffin) | |
| V9 | |
| Unconjugated | |
| Purified vimentin from pig eye lens | |
| IVD | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.