missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Villin Mouse anti-Human, Mouse, Clone: 2B7B9, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | Villin |
|---|---|
| Clone | 2B7B9 |
| Concentration | 0.89 mg/mL |
| Applications | Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16807665
|
Proteintech
66096-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16897655
|
Proteintech
66096-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Villin is part of cytoskeleton associated with the microvillar actin core bundle of gastrointestinal and renal brush border. It can interact with actin in a Ca2+ and phosphoinositide-regulated manner. Villin is a very specific marker for adenocarcinomas of the gastrointestinal tract and the pancreas. It is also expressed in some Merkel cell tumor and adenocarcinoma of the lung, prostate, ovarian and kidney.Specifications
| Villin | |
| 0.89 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| VIL, VIL1, Villin, villin 1 | |
| VIL1 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 2B7B9 | |
| Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| P09327, Q62468 | |
| 22349, 7429 | |
| Villin Fusion Protein Ag9637 | |
| Primary | |
| -20°C | |
| VIL1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title