missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Villin Mouse anti-Human, Mouse, Clone: 2B7B9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66096-1-IG-150UL
This item is not returnable.
View return policy
Description
Villin is part of cytoskeleton associated with the microvillar actin core bundle of gastrointestinal and renal brush border. It can interact with actin in a Ca2+ and phosphoinositide-regulated manner. Villin is a very specific marker for adenocarcinomas of the gastrointestinal tract and the pancreas. It is also expressed in some Merkel cell tumor and adenocarcinoma of the lung, prostate, ovarian and kidney.Specifications
| Villin | |
| Monoclonal | |
| 0.89 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P09327, Q62468 | |
| VIL1 | |
| Villin Fusion Protein Ag9637 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 2B7B9 | |
| Unconjugated | |
| VIL1 | |
| VIL, VIL1, Villin, villin 1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 22349, 7429 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction