missing translation for 'onlineSavingsMsg'
Learn More
Learn More
villin-like Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | villin-like |
|---|---|
| Dilution | Western Blot 1:200 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18693770
|
Novus Biologicals
NBP2-93027-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18601681
|
Novus Biologicals
NBP2-93027-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
villin-like Polyclonal antibody specifically detects villin-like in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| villin-like | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 50853 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.2.1.2, EC 6.3.2.6, VILL, villin like, Villin-Like, Villin-Like Protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 637-856 of human VILL (NP_056957.3). LDKYDIMLLDTWQEIFLWLGEAASEWKEAVAWGQEYLKTHPAGRSPATPIVLVKQGHEPPTFIGWFFTWDPYKWTSHPSHKEVVDGSPAAASTISEITAEVNNLRLSRWPGNGRAGAVALQALKGSQDSSENDLVRSPKSAGSRTSSSVSSTSATINGGLRREQLMHQAVEDLPEGVDPARREFYLSDSDFQDIFGKSKEEFYSMATWRQRQEKKQLGFF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title