missing translation for 'onlineSavingsMsg'
Learn More
Learn More
villin-like Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93027-0.1ml
This item is not returnable.
View return policy
Description
villin-like Polyclonal antibody specifically detects villin-like in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| villin-like | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| EC 2.2.1.2, EC 6.3.2.6, VILL, villin like, Villin-Like, Villin-Like Protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 637-856 of human VILL (NP_056957.3). LDKYDIMLLDTWQEIFLWLGEAASEWKEAVAWGQEYLKTHPAGRSPATPIVLVKQGHEPPTFIGWFFTWDPYKWTSHPSHKEVVDGSPAAASTISEITAEVNNLRLSRWPGNGRAGAVALQALKGSQDSSENDLVRSPKSAGSRTSSSVSSTSATINGGLRREQLMHQAVEDLPEGVDPARREFYLSDSDFQDIFGKSKEEFYSMATWRQRQEKKQLGFF | |
| 0.1 mL | |
| Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 50853 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction