missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VILIP 3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | VILIP 3 |
|---|---|
| Concentration | 0.28 mg/mL |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16867743
|
Proteintech
10989-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16857743
|
Proteintech
10989-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hippocalcin-like protein 1, may be involved in the calcium-dependent regulation of rhodopsin phosphorylation.Specifications
| VILIP 3 | |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P37235, P62748 | |
| 3241, 53602 | |
| VILIP 3 Fusion Protein Ag1449 | |
| Primary | |
| -20°C | |
| HPCAL1 |
| 0.28 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| BDR1, Calcium binding protein BDR 1, hippocalcin like 1, Hippocalcin like protein 1, HLP2, HPCA, HPCAL1, VILIP 3, Visinin like protein 3 | |
| HPCAL1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title