missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VILIP 3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10989-1-AP-150UL
This item is not returnable.
View return policy
Description
Hippocalcin-like protein 1, may be involved in the calcium-dependent regulation of rhodopsin phosphorylation.Specifications
| VILIP 3 | |
| Polyclonal | |
| Unconjugated | |
| HPCAL1 | |
| BDR1, Calcium binding protein BDR 1, hippocalcin like 1, Hippocalcin like protein 1, HLP2, HPCA, HPCAL1, VILIP 3, Visinin like protein 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3241, 53602 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P37235, P62748 | |
| HPCAL1 | |
| VILIP 3 Fusion Protein Ag1449 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction