Learn More
Invitrogen™ FGF12 Monoclonal Antibody (K94041_8C12), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555999
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: KGIVTRLFSQ QGYFLQMHPD GTIDGTKDEN SDYTLFNLIP VGLRVVAIQG VKASLYVAMN GEGYLYSSDV FTPECKFKES VFENYYVIYS STLYRQQESG RAWFLGLNKE GQIMKGNRVK KTKPSSHFVP KPIEVCMYRE PSLHEIGEKQ GRSRKSSGTP TMNGGKVVNQ
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth, and invasion. This growth factor lacks the N-terminal signal sequence present in most of the FGF family members, but it contains clusters of basic residues that have been demonstrated to act as a nuclear localization signal. When transfected into mammalian cells, this protein accumulated in the nucleus, but was not secreted. The specific function of this gene has not yet been determined. Two alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
| FGF12 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P61150, P61328, P61329 | |
| Fgf12 | |
| Recombinant human FGF12 (immunogen range is 71-240). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot | |
| K94041_8C12 | |
| Unconjugated | |
| Fgf12 | |
| AV114868; B230343J05Rik; FGF; FGF12; FGF-12; FGF12B; FHF1; FHF-1; Fhf1b; Fibroblast growth factor; Fibroblast growth factor 12; fibroblast growth factor 12B; fibroblast growth factor FGF-12b; fibroblast growth factor homologous factor 1; fibroblast growth factor homologous factor 1b; myocyte-activating factor | |
| Mouse | |
| Protein A | |
| RUO | |
| 14167, 170630, 2257 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |