missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FGF12 Monoclonal Antibody (K94041_8C12), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | FGF12 |
|---|---|
| Clone | K94041_8C12 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30199806
|
Invitrogen™
MA555999 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30203660
|
Invitrogen™
MA555619 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: KGIVTRLFSQ QGYFLQMHPD GTIDGTKDEN SDYTLFNLIP VGLRVVAIQG VKASLYVAMN GEGYLYSSDV FTPECKFKES VFENYYVIYS STLYRQQESG RAWFLGLNKE GQIMKGNRVK KTKPSSHFVP KPIEVCMYRE PSLHEIGEKQ GRSRKSSGTP TMNGGKVVNQ
FGF12 Monoclonal antibody specifically detects FGF12 in Human, Mouse, Rat samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| FGF12 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| AV114868; B230343J05Rik; FGF; FGF12; FGF-12; FGF12B; FHF1; FHF-1; Fhf1b; Fibroblast growth factor; Fibroblast growth factor 12; fibroblast growth factor 12B; fibroblast growth factor FGF-12b; fibroblast growth factor homologous factor 1; fibroblast growth factor homologous factor 1b; myocyte-activating factor | |
| Recombinant human FGF12 (immunogen range is 71-240). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Fgf12 |
| K94041_8C12 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| P61150, P61328, P61329 | |
| 14167, 170630, 2257 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title