Learn More
Human/Mouse Dkk-1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 15 publications
Marca: R&D Systems AF1096-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Produced in goats immunized with purified, insect cell line Sf 21-derived, recombinant human Dickkopf homolog-1 (rhDkk-1). Human Dkk-1 specific IgG was purified by human Dkk-1 affinity chromatography.
Dkk-1 Polyclonal specifically detects Dkk-1 in Human, Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction, Knockout Validated.
Especificaciones
| Dkk-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative | |
| dickkopf (Xenopus laevis) homolog 1, dickkopf homolog 1 (Xenopus laevis), dickkopf related protein-1, Dickkopf-1, dickkopf-related protein 1, Dkk1, DKK-1, hDkk-1, SKdickkopf-1 like | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 22943 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Blockade of Receptor-ligand Interaction, Immunoassay | |
| LYOPH | |
| Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction, Knockout Validated | |
| O94907 | |
| DKK1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Dkk-1 Thr32-His266 Accession # O94907 | |
| 25 μg | |
| Primary | |
| Detects human and mouse Dkk-1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) Dkk-2, and rhDkk-4 is observed. | |
| Human, Mouse | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido