missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Dkk-1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 15 publications
£187.00 - £512.00
Specifications
| Antigen | Dkk-1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction, Knockout Validated |
| Applications | Western Blot, Blockade of Receptor-ligand Interaction, Immunoassay |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18799067
![]() |
R&D Systems
AF1096-SP |
25 μg |
£187.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18001762
|
R&D Systems
AF1096 |
100 μg |
£512.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Produced in goats immunized with purified, insect cell line Sf 21-derived, recombinant human Dickkopf homolog-1 (rhDkk-1). Human Dkk-1 specific IgG was purified by human Dkk-1 affinity chromatography.
Dkk-1 Polyclonal specifically detects Dkk-1 in Human, Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction, Knockout Validated.Specifications
| Dkk-1 | |
| Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction, Knockout Validated | |
| Polyclonal | |
| Goat | |
| Human, Mouse | |
| dickkopf (Xenopus laevis) homolog 1, dickkopf homolog 1 (Xenopus laevis), dickkopf related protein-1, Dickkopf-1, dickkopf-related protein 1, Dkk1, DKK-1, hDkk-1, SKdickkopf-1 like | |
| DKK1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Blockade of Receptor-ligand Interaction, Immunoassay | |
| Unconjugated | |
| RUO | |
| O94907 | |
| 22943 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Dkk-1 Thr32-His266 Accession # O94907 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human and mouse Dkk-1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) Dkk-2, and rhDkk-4 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title