Learn More
Human Phospho-STAT5a/b (Y694/Y699) Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  R&D Systems MAB4190
 View more versions of this product
Product Code. 18785358
  • £422.00 / 100µg
Description
STAT5a/b Monoclonal specifically detects STAT5a/b in Human samples. It is validated for Western Blot, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
STAT5a/b
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
MGF, STAT5, STAT5A signal transducer and activator of transcription 5A
Phosphopeptide containing human STAT5b Y699 site (amino acid sequence of this peptide is identical to a corresponding region of human STAT5a containing Y694). Accession # P51692
100 μg
Primary
Detects human STAT5a/b when phosphorylated at Y694/Y699 in Western blots.
Human
Purified
Western Blot, Immunocytochemistry
941212
Western Blot 2 ug/mL, Immunocytochemistry 0.3-25 ug/mL
P51692
Rat
Protein A or G purified from cell culture supernatant
RUO
6776
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a