missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-STAT5a/b (Y694/Y699) Antibody, R&D Systems™

Rat Monoclonal Antibody
£168.00 - £422.00
Specifications
| Antigen | STAT5a/b |
|---|---|
| Clone | 941212 |
| Dilution | Western Blot 2 ug/mL, Immunocytochemistry 0.3-25 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18774258
![]() |
R&D Systems
MAB4190-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18785358
![]() |
R&D Systems
MAB4190 |
100 μg |
£422.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
STAT5a/b Monoclonal specifically detects STAT5a/b in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| STAT5a/b | |
| Western Blot 2 ug/mL, Immunocytochemistry 0.3-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MGF, STAT5, STAT5A signal transducer and activator of transcription 5A | |
| Phosphopeptide containing human STAT5b Y699 site (amino acid sequence of this peptide is identical to a corresponding region of human STAT5a containing Y694). Accession # P51692 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human STAT5a/b when phosphorylated at Y694/Y699 in Western blots. |
| 941212 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rat | |
| Human | |
| P51692 | |
| 6776 | |
| IgG2a | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title